missing translation for 'onlineSavingsMsg'
Learn More

S-Methylisothiourea Sulfate 98.0+%, TCI America™

Catalog No. M044225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044225G 25 g
M0442100G 100 g
M0442500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M044225G Supplier TCI America Supplier No. M044225G

Chemical Identifiers

CAS 867-44-7
Molecular Formula C4H14N4O4S3
Molecular Weight (g/mol) 278.36
MDL Number MFCD00013055
InChI Key BZZXQZOBAUXLHZ-UHFFFAOYSA-N
Synonym s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
PubChem CID 13347
IUPAC Name methyl carbamimidothioate;sulfuric acid
SMILES CSC(=N)N.CSC(=N)N.OS(=O)(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 278.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-Methylisothiourea Sulfate
EINECSNumber (2)-2865&(9)-1413
RTECSNumber UM9200000
TSCA Yes