missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl Thiobutyrate 98.0+%, TCI America™

Catalog No. M181825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M181825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M181825G Supplier TCI America Supplier No. M181825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2432-51-1
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00009872
InChI Key GRLJIIJNZJVMGP-UHFFFAOYSA-N
Synonym s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
PubChem CID 62444
IUPAC Name S-methyl butanethioate
SMILES CCCC(=O)SC

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 g
UN Number 1993
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Methyl Thiobutyrate