missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Methyl Methanethiosulfonate 97.0+%, TCI America™
Supplier: TCI America M138225G
Specifications
| S-Methyl Methanethiosulfonate | |
| 125°C | |
| MFCD00007565 | |
| 2810 | |
| XYONNSVDNIRXKZ-UHFFFAOYSA-N | |
| methylsulfonylsulfanylmethane | |
| 18064 | |
| 126.19 | |
| Liquid |
| 2949-92-0 | |
| C2H6O2S2 | |
| 25 g | |
| s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane | |
| CSS(=O)(=O)C | |
| 126.188 | |
| CHEBI:74357 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2949-92-0 | |
| 126.188 | |
| XYONNSVDNIRXKZ-UHFFFAOYSA-N | |
| 18064 | |
| methylsulfonylsulfanylmethane |
| C2H6O2S2 | |
| MFCD00007565 | |
| s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane | |
| CHEBI:74357 | |
| CSS(=O)(=O)C |
Safety and Handling
RTECSNumber : PB2765000
TSCA : Yes