missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl Methanethiosulfonate 97.0+%, TCI America™

Catalog No. p-7132697
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13825G 5 g
M138225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13825G Supplier TCI America Supplier No. M13825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2949-92-0
Molecular Formula C2H6O2S2
Molecular Weight (g/mol) 126.188
MDL Number MFCD00007565
InChI Key XYONNSVDNIRXKZ-UHFFFAOYSA-N
Synonym s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane
PubChem CID 18064
ChEBI CHEBI:74357
IUPAC Name methylsulfonylsulfanylmethane
SMILES CSS(=O)(=O)C

Specifications

Boiling Point 125°C
Quantity 5 g
UN Number 2810
Formula Weight 126.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Methyl Methanethiosulfonate
RTECSNumber PB2765000
TSCA Yes