accessibility menu, dialog, popup

UserName

CAS RN 2949-92-0

IUPAC Name: (methanesulfonylsulfanyl)methane
Synonyms: methyl methanethiosulfonate
Molecular Weight (g/mol): 126.19
Molecular Formula: C2H6O2S2
InChi Key: XYONNSVDNIRXKZ-UHFFFAOYSA-N
SMILES: CSS(C)(=O)=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)