missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Methyl-L-cysteine 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 135.18 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | S-Methyl-L-cysteine |
Safety and Handling
| RTECSNumber | AY7175000 |
| TSCA | No |