missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl 2-Furancarbothioate 98.0+%, TCI America™

Catalog No. M182025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M182025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M182025G Supplier TCI America Supplier No. M182025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13679-61-3
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD00040266
InChI Key ISKUAGFDTRLBHG-UHFFFAOYSA-N
Synonym 2-Furancarbothioic Acid S-Methyl Ester, S-Methyl 2-Thiofuroate, 2-Thiofuroic Acid S-Methyl Ester
PubChem CID 61662
IUPAC Name S-methyl furan-2-carbothioate
SMILES CSC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 63°C
Quantity 25 g
Formula Weight 142.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Methyl 2-Furancarbothioate