missing translation for 'onlineSavingsMsg'
Learn More

s-Diphenylcarbazone, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCD069010F
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCD069010F 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCD069010F Supplier Ricca Chemical Company Supplier No. RDCD069010F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 538-62-5 , 140-22-7
Molecular Formula C13H12N4O
Molecular Weight (g/mol) 240.266
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
PubChem CID 10860
IUPAC Name 1-anilino-3-phenyliminourea
SMILES C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Orange
Packaging Pouch
Quantity 10 g
Grade ACS
Physical Form Powder
Chemical Name or Material s-Diphenylcarbazone
ShelfLife 24 months