missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-DBD-Pro-COCl 90,0+%, TCI America™

Numéro de catalogue. A5564100MG
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A5564100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A5564100MG Fournisseur TCI America Code fournisseur A5564100MG

[=(S)-(-)-4-(N,N-Diméthylaminosulfonyl)-7-(2-chloroformylpyrrolidine-1-yl)-2,1,3-benzoxadiazole], [Réactif de marquage HPLC pour la détermination, c.-é.]

Identifiants chimiques

CAS 150993-63-8
Molecular Formula C13H15ClN4O4S
Molecular Weight (g/mol) 358.797
MDL Number MFCD00191380
InChI Key ZPBKQTOHYMFDLD-VIFPVBQESA-N
Synonym (S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 44630074
IUPAC Name (2S)-1-[4-(diméthylsulfamoyl)-2,1,3-benzoxadiazol-7-yl]pyrrolidine-2-chlorure de carbonyle
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N3CCCC3C(=O)Cl

Spécifications

Color Jaune
Quantity 100 mg
Formula Weight 358.80
Percent Purity ≥90.0% (HPLC)
Physical Form Poudre cristalline
Chemical Name or Material (S)-(-)-DBD-Pro-COCl [=(S)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl
TSCA No
Recommended Storage Réfrigérateur