missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Carvone 98.0+%, TCI America™

Catalog No. C070425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C070425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C070425ML Supplier TCI America Supplier No. C070425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2244-16-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00062997
InChI Key ULDHMXUKGWMISQ-VIFPVBQESA-N
Synonym d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
PubChem CID 16724
ChEBI CHEBI:15399
IUPAC Name (5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC1=CCC(CC1=O)C(=C)C

Specifications

Boiling Point 230°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-Carvone
EINECSNumber (3)-2402
RTECSNumber OS8670000
TSCA Yes