missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Carvone, 96%

Numéro de catalogue. p-7044564
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0713014 25 g
AAL0713022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0713014 Fournisseur Thermo Scientific Chemicals Code fournisseur L0713014
Il en reste null

CAS: 2244-16-8 | C10H14O | 150.221 g/mol

Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

Solubility
Miscible with diethyl ether, ethanol and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents. Keep the container tightly closed in a dry and well-ventilated place.

Identifiants chimiques

CAS 2244-16-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00062997
InChI Key ULDHMXUKGWMISQ-VIFPVBQESA-N
Synonym d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
PubChem CID 16724
ChEBI CHEBI:15399
IUPAC Name (5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC1=CCC(CC1=O)C(=C)C

Spécifications

Density 0.959
Boiling Point 228°C to 230°C
Flash Point 93°C (199°F)
Odor Pleasant
Refractive Index 1.498
Quantity 25 g
Beilstein 2042970
Merck Index 14,1874
Solubility Information Miscible with diethyl ether,ethanol and chloroform. Immiscible with water.
Optical Rotation +59° (Neat)
Formula Weight 150.22
Percent Purity 96%
Chemical Name or Material (S)-(+)-Carvone
Afficher plus de résultats Afficher moins de résultats
Hazard Category H317
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 218-827-2
RTECSNumber OS8670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only