missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Carvone, 96%

Catalog No. AAL0713022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0713022 100 g
AAL0713014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0713022 Supplier Thermo Scientific Chemicals Supplier No. L0713022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2244-16-8 | C10H14O | 150.221 g/mol

Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

Solubility
Miscible with diethyl ether, ethanol and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 2244-16-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00062997
InChI Key ULDHMXUKGWMISQ-VIFPVBQESA-N
Synonym d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
PubChem CID 16724
ChEBI CHEBI:15399
IUPAC Name (5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC1=CCC(CC1=O)C(=C)C

Specifications

Density 0.959
Boiling Point 228°C to 230°C
Flash Point 93°C (199°F)
Odor Pleasant
Refractive Index 1.498
Quantity 100 g
Beilstein 2042970
Merck Index 14,1874
Solubility Information Miscible with diethyl ether,ethanol and chloroform. Immiscible with water.
Optical Rotation +59° (Neat)
Formula Weight 150.22
Percent Purity 96%
Chemical Name or Material (S)-(+)-Carvone
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 218-827-2
RTECSNumber OS8670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only