missing translation for 'onlineSavingsMsg'
Learn More

S-Butyl Thiobenzoate 97.0+%, TCI America™

Catalog No. B100925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B100925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B100925ML Supplier TCI America Supplier No. B100925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7269-35-4
Molecular Formula C11H14OS
Molecular Weight (g/mol) 194.292
MDL Number MFCD00039955
InChI Key KXXZPIFZTPWOJJ-UHFFFAOYSA-N
Synonym Thiobenzoic Acid S-Butyl Ester
PubChem CID 12373904
IUPAC Name S-butyl benzenecarbothioate
SMILES CCCCSC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 194.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Butyl Thiobenzoate