missing translation for 'onlineSavingsMsg'
Learn More

S-Benzylisothiourea Hydrochloride 99.0+%, TCI America™

Catalog No. B044225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B044225G Supplier TCI America Supplier No. B044225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 538-28-3
Molecular Formula C8H11ClN2S
Molecular Weight (g/mol) 202.7
MDL Number MFCD00012583
InChI Key WJAASTDRAAMYNK-UHFFFAOYSA-N
Synonym s-benzylisothiourea hydrochloride, 2-benzyl-2-thiopseudourea hydrochloride, btkh, s-benzylthiuronium chloride, benzyl carbamimidothioate hydrochloride, s-benzylthiouronium chloride, benzylisothiourea hydrochloride, benzylthiuronium chloride, 2-benzylisothiouronium chloride, benzylisothiuronium chloride
PubChem CID 10852
IUPAC Name benzyl carbamimidothioate;hydrochloride
SMILES C1=CC=C(C=C1)CSC(=N)N.Cl

Specifications

Melting Point 176°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 202.70
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-Benzylisothiourea Hydrochloride
RTECSNumber UM0738000
TSCA Yes