missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. M08295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08295G 5 g
M08291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08295G Supplier TCI America Supplier No. M08295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26164-26-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00064216
InChI Key DIWVBIXQCNRCFE-QMMMGPOBSA-N
Synonym s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
PubChem CID 643325
IUPAC Name (2S)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-alpha-Methoxyphenylacetic Acid