missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride, TCI America™

Catalog No. M22145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22145G Supplier TCI America Supplier No. M22145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(ca. 18% in Dichloromethane, ca. 1.0mol/L), [for Determination of the optical purity of Alcohols and Amines]

Chemical Identifiers

CAS 20445-33-4
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.617
MDL Number MFCD00067105
InChI Key PAORVUMOXXAMPL-SECBINFHSA-N
Synonym s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride
PubChem CID 2724611
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 252.62
Physical Form Liquid
Chemical Name or Material (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl Chloride (ca. 18% in Dichloromethane,
TSCA No
Recommended Storage Refrigerator