missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%

CAS: 17257-71-5 | C10H8F3O3 | 233.17 g/mol

Supplier:  Thermo Scientific Chemicals B2311788

 View more versions of this product

Catalog No. AAB2311788


Only null left
Explore available promotions

Description

Description

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is used as a chiral derivatizing agent, it reacts with an alcohol or amine of unknown stereochemistry to form an ester or amide. The absolute configuration of the ester or amide is then determined by proton and/or 19F NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is used as a chiral derivatizing agent, it reacts with an alcohol or amine of unknown stereochemistry to form an ester or amide. The absolute configuration of the ester or amide is then determined by proton and/or 19F NMR spectroscopy.

Solubility
Readily soluble in hexane, ether, THF, CH2Cl2, benzene.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications

Specifications

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid
43°C to 45°C
105°C to 107°C (1 mmHg)
C10H8F3O3
2.5 g
Hygroscopic
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
JJYKJUXBWFATTE-VIFPVBQESA-M
CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F
233.17
234.18
17257-71-5
1.344
>110°C (230°F)
MFCD00064200
3591561
14,6280
Readily soluble in hexane,ether,THF,CH2Cl2,benzene.
−72° (c=2 in Methanol)
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
6992788
99%

Chemical Identifiers

17257-71-5
233.17
JJYKJUXBWFATTE-VIFPVBQESA-M
6992788
CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F
C10H8F3O3
MFCD00064200
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-292-1

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only