accessibility menu, dialog, popup

UserName

H3CO(S)OOFFF

CAS RN 17257-71-5

IUPAC Name: (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
Synonyms: (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
Molecular Weight (g/mol): 233.17
Molecular Formula: C10H8F3O3
InChi Key: JJYKJUXBWFATTE-VIFPVBQESA-M
SMILES: CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)