missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha,alpha-Diphenylprolinol, 98%, Thermo Scientific Chemicals

Catalog No. AAL0921703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0921703 1 g
AAL0921706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0921703 Supplier Thermo Scientific Chemicals Supplier No. L0921703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-alpha,alpha-Diphenylprolinol is used to prepare the corresponding oxazaborolidines for the borane-mediated asymmetric reduction of ketones.

Solubility
Soluble in chloroform.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 112068-01-6
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.345
MDL Number MFCD00075506
InChI Key OGCGXUGBDJGFFY-INIZCTEOSA-N
Synonym s---alpha,alpha-diphenyl-2-pyrrolidinemethanol, s-diphenyl pyrrolidin-2-yl methanol, s---2-diphenylhydroxymethyl pyrrolidine, alpha,alpha-diphenyl-l-prolinol, diphenyl-l-pyrrolidin-2-yl-methanol, diphenyl-2s-pyrrolidin-2-yl methanol, s-alpha,alpha-diphenyl-2-pyrrolidinemethanol, s---diphenylpyrrolidinemethanol, s---alpha,alpha-diphenyl-2-pyrrolidine methanol, diphenyl 2s-pyrrolidin-2-ylmethanol
PubChem CID 2724899
IUPAC Name diphenyl-[(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 76°C to 78°C
Quantity 1 g
Optical Rotation −57.5° (c=3 in Methanol)
Beilstein 17103
Solubility Information Soluble in chloroform.
Formula Weight 253.35
Percent Purity 98%
Chemical Name or Material (S)-(-)-alpha,alpha-Diphenylprolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only