Learn More
Electron Microscopy Sciences S-52 Sedgewick Rafter, Glass
Numéro de catalogue. 5028541
Encompass
Used to create precise volumes of liquid to enable counting of particles in a specified volume of liquid. For use in water analysis, culture inspection, and any fluid where particles per unit volume need to be determined. Each cell is supplied with one cover glass. Made from glass, with a chromium surface image. For professional use and when using phase contrast. With one cover glass.
Numéro de catalogue. 5028541 Fournisseur Electron Microscopy Sciences Code fournisseur 6805052
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Used to create precise volumes of liquid to enable counting of particles in a specified volume of liquid. For use in water analysis, culture inspection, and any fluid where particles per unit volume need to be determined. Each cell is supplied with one cover glass. Made from glass, with a chromium surface image. For professional use and when using phase contrast. With one cover glass.