missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-4-Phenyl-2-oxazolidinone 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 131°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 163.18 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (S)-(+)-4-Phenyl-2-oxazolidinone |
Safety and Handling
| TSCA | No |