missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Isopropylthiazolidine-2-thione 98.0+%, TCI America™

Catalog No. I05751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05751G Supplier TCI America Supplier No. I05751G
Only null left

Chemical Identifiers

CAS 76186-04-4
Molecular Formula C6H11NS2
Molecular Weight (g/mol) 161.281
MDL Number MFCD03844797
InChI Key CWIZUGZKLJDJLE-RXMQYKEDSA-N
Synonym (S)-4,5-Dihydro-4-isopropylthiazole-2-thiol
PubChem CID 11321340
IUPAC Name (4S)-4-propan-2-yl-1,3-thiazolidine-2-thione
SMILES CC(C)C1CSC(=S)N1

Specifications

Melting Point 73°C
Color White
Quantity 1 g
Formula Weight 161.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-Isopropylthiazolidine-2-thione