missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane 97.0+%, TCI America™

Catalog No. p-7125175
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17961G 1 g
C17965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17961G Supplier TCI America Supplier No. C17961G
Only null left

Chemical Identifiers

CAS 60456-22-6
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00273365
InChI Key BNPOTXLWPZOESZ-RXMQYKEDSA-N
Synonym s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa
PubChem CID 6930483
IUPAC Name (4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
SMILES CC1(OCC(O1)CCl)C

Specifications

Color Colorless
Boiling Point 63°C
Quantity 1 g
UN Number 1992
Formula Weight 150.60
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
RTECSNumber JH6950000
TSCA No