missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. B21371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21371G Supplier TCI America Supplier No. B21371G
Only null left

Chemical Identifiers

CAS 16495-03-7
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD00043000
InChI Key DBFDSKSLTCMIPB-LBPRGKRZSA-N
PubChem CID 10878747
IUPAC Name (4S)-2,2-dimethyl-4-(phenylmethoxymethyl)-1,3-dioxolane
SMILES CC1(OCC(O1)COCC2=CC=CC=C2)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 222.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator