missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Benzyl-2-oxazolidinone, 99%

Catalog No. p-7024061
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1823606 5 g
AAA1823614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1823606 Supplier Thermo Scientific Chemicals Supplier No. A1823606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90719-32-7 | C10H11NO2 | 177.20 g/mol

(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 90719-32-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00064496
InChI Key OJOFMLDBXPDXLQ-VIFPVBQESA-N
Synonym s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
PubChem CID 736225
IUPAC Name (4S)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 87°C to 89°C
Beilstein 3649667
Solubility Information Insoluble in water.
Quantity 5 g
Optical Rotation −64° (c=1 in Chloroform)
Formula Weight 177.2
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material (S)-(-)-4-Benzyl-2-oxazolidinone
Hazard Category H302-H315-H320-H335-H373
Precautionary Statement P261-P262-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P314-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only