missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-4-Amino-2-hydroxybutyric Acid 98.0+%, TCI America™
$47.46 - $146.20
Chemical Identifiers
| CAS | 40371-51-5 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00674110 |
| InChI Key | IVUOMFWNDGNLBJ-VKHMYHEASA-N |
| Synonym | s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid |
| PubChem CID | 2733929 |
| IUPAC Name | (2S)-4-azaniumyl-2-hydroxybutanoate |
| SMILES | [NH3+]CC[C@H](O)C([O-])=O |
Chemical Identifiers
| 40371-51-5 | |
| 119.12 | |
| IVUOMFWNDGNLBJ-VKHMYHEASA-N | |
| 2733929 | |
| [NH3+]CC[C@H](O)C([O-])=O |
| C4H9NO3 | |
| MFCD00674110 | |
| s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid | |
| (2S)-4-azaniumyl-2-hydroxybutanoate |
Specifications
| 203°C | |
| White | |
| MFCD00674110 | |
| IVUOMFWNDGNLBJ-VKHMYHEASA-N | |
| (2S)-4-azaniumyl-2-hydroxybutanoate | |
| 2733929 | |
| ≥98.0% (T) | |
| (S)-(-)-4-Amino-2-hydroxybutyric Acid |
| 40371-51-5 | |
| C4H9NO3 | |
| s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid | |
| [NH3+]CC[C@H](O)C([O-])=O | |
| 119.12 | |
| 119.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No