accessibility menu, dialog, popup

UserName

CAS RN 40371-51-5

IUPAC Name: (2S)-4-azaniumyl-2-hydroxybutanoate
Synonyms: (2S)-4-ammonio-2-hydroxybutanoate
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: IVUOMFWNDGNLBJ-VKHMYHEASA-N
SMILES: [NH3+]CC[C@H](O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)