missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Amino-2-hydroxybutyric Acid 98.0+%, TCI America™

Catalog No. A14385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14385G 5 g
A143825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14385G Supplier TCI America Supplier No. A14385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40371-51-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00674110
InChI Key IVUOMFWNDGNLBJ-VKHMYHEASA-N
Synonym s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid
PubChem CID 2733929
IUPAC Name (2S)-4-azaniumyl-2-hydroxybutanoate
SMILES [NH3+]CC[C@H](O)C([O-])=O

Specifications

Melting Point 203°C
Color White
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-4-Amino-2-hydroxybutyric Acid