missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Amino-2-hydroxybutyric Acid 98.0+%, TCI America™

Supplier:  TCI America A143825G

Quantity 25 g

 View more versions of this product

Catalog No. A143825G


Only null left

Description

Description

Specifications

Chemical Identifiers

40371-51-5
119.12
IVUOMFWNDGNLBJ-VKHMYHEASA-N
2733929
[NH3+]CC[C@H](O)C([O-])=O
C4H9NO3
MFCD00674110
s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid
(2S)-4-azaniumyl-2-hydroxybutanoate

Specifications

203°C
40371-51-5
C4H9NO3
s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid
[NH3+]CC[C@H](O)C([O-])=O
25 g
2733929
≥98.0% (T)
(S)-(-)-4-Amino-2-hydroxybutyric Acid
White
MFCD00674110
IVUOMFWNDGNLBJ-VKHMYHEASA-N
(2S)-4-azaniumyl-2-hydroxybutanoate
119.12
119.12
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions