missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Methylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™

Catalog No. M21575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21575G 5 g
M215725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21575G Supplier TCI America Supplier No. M21575G
Only null left

Chemical Identifiers

CAS 116539-55-0
Molecular Formula C8H13NOS
Molecular Weight (g/mol) 171.258
MDL Number MFCD07357308
InChI Key YEJVVFOJMOHFRL-ZETCQYMHSA-N
Synonym (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene
PubChem CID 10095047
IUPAC Name (1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol
SMILES CNCCC(C1=CC=CS1)O

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 171.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(Methylamino)-1-(2-thienyl)-1-propanol