missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%

Catalog No. AAL19041MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19041MD 250 mg
AAL1904103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19041MD Supplier Thermo Scientific Chemicals Supplier No. L19041MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36567-72-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36567-72-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00145262
InChI Key AYOLELPCNDVZKZ-QMMMGPOBSA-N
Synonym s-3-hydroxy-3-phenylpropanoic acid, 3s-3-hydroxy-3-phenylpropanoic acid, s-3-hydroxy-3-phenylpropionic acid, ?-3-phenylhydracrylic acid, s---3-hydroxy-3-phenylpropionic acid, benzenepropanoic acid,, a-hydroxy-,, as, s---3-hydroxy-3-phenylpropionic acid, 98+%, unii-s2w4lws09c component, s-3-hydroxy-3-phenylpropionic acid sum of enantiomers, gc
PubChem CID 2735057
ChEBI CHEBI:51058
IUPAC Name (3S)-3-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)O

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
Beilstein 3197773
Optical Rotation −22° (c=4 in Methanol)
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-3-Hydroxy-3-phenylpropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only