missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%

Catalog No. p-7030010
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27536MD 250 mg
AAH2753603 1 g
AAH2753606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH27536MD Supplier Thermo Scientific Chemicals Supplier No. H27536MD

CAS: 136725-53-6 | C4H9ClFN | 125.571 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136725-53-6
Molecular Formula C4H9ClFN
Molecular Weight (g/mol) 125.571
MDL Number MFCD04038718
InChI Key LENYOXXELREKGZ-WCCKRBBISA-N
Synonym s-3-fluoropyrrolidine hydrochloride, s-+-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine hydrochloride, 3s-+-3-fluoropyrrolidine hydrochloride, s-3-fluoro-pyrrolidine hcl, s ;-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine, chloride, s-3-fluoropyrrolidine hcl
PubChem CID 16217739
IUPAC Name (3S)-3-fluoropyrrolidine;hydrochloride
SMILES C1CNCC1F.Cl

Specifications

Melting Point 183°C to 186°C
Quantity 250 mg
Formula Weight 125.57
Percent Purity 97%
Chemical Name or Material (S)-(+)-3-Fluoropyrrolidine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only