missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

Catalog No. AAH5198706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5198706 5 g
AAH5198703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5198706 Supplier Thermo Scientific Chemicals Supplier No. H5198706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162558-25-0 | C23H26N2O6 | 426.469 g/mol

As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 162558-25-0
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00235896
InChI Key PKAUMAVONPSDRW-IBGZPJMESA-N
Synonym fmoc-dap boc-oh, n-fmoc-n'-boc-l-2,3-diaminopropionic acid, fmoc-dpr boc-oh, fmoc-l-2,3-diaminopropionic acid boc, fmoc-alpha,beta-diaminopropionic acid boc-oh, 2s-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-dap boc, fmoc-dap boc, fmoc-l-dpa boc-oh, ambotzfaa1367
PubChem CID 2756103
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 5 g
Beilstein 7057792
Solubility Information Slightly soluble in water.
Formula Weight 426.47
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only