missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-3-Benzyloxy-1,2-propanediol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | White-Yellow |
| Quantity | 100 mg |
| Formula Weight | 182.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | (S)-(-)-3-Benzyloxy-1,2-propanediol |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |