missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-1,2-propanediol 98.0+%, TCI America™

Catalog No. A19805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19805G 5 g
A19801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19805G Supplier TCI America Supplier No. A19805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61278-21-5
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00798260
InChI Key KQIGMPWTAHJUMN-VKHMYHEASA-N
Synonym s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
PubChem CID 440902
ChEBI CHEBI:1456
IUPAC Name (2S)-3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
UN Number 3259
Formula Weight 91.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-3-Amino-1,2-propanediol
EINECSNumber (2)-3504
RTECSNumber TY2800000
TSCA No