missing translation for 'onlineSavingsMsg'
Learn More

(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid 98.0+%, TCI America™

Catalog No. T19011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19011G Supplier TCI America Supplier No. T19011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24435-45-8
Molecular Formula C4H5F3O3
Molecular Weight (g/mol) 158.076
MDL Number MFCD03092898
InChI Key CTGJACFEVDCYMC-VKHMYHEASA-N
Synonym (S)-2-Hydroxy-2-(trifluoromethyl)propionic Acid, (S)-3,3,3-Trifluoro-2-hydroxyisobutyric Acid
PubChem CID 7019028
IUPAC Name (2S)-3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
SMILES CC(C(=O)O)(C(F)(F)F)O

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 158.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid
TSCA No
Recommended Storage Refrigerator