missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(tert-Butoxycarbonylamino)butyric Acid 98.0+%, TCI America™

Catalog No. B51155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51155G 5 g
B511525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51155G Supplier TCI America Supplier No. B51155G

Chemical Identifiers

CAS 34306-42-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037267
InChI Key PNFVIPIQXAIUAY-UHFFFAOYNA-N
Synonym boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
PubChem CID 2755934
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}butanoic acid
SMILES CCC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 75°C
Color White
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(tert-Butoxycarbonylamino)butyric Acid