missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%

Catalog No. AAL1171136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1171136 500 g
AAL1171114 25 g
AAL1171122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1171136 Supplier Thermo Scientific Chemicals Supplier No. L1171136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-79-3 | C5H7NO3 | 129.12 g/mol

(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.

Solubility
Soluble in water (10-15% at 20°C), alcohol, acetone, dimethyl sufoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
PubChem CID 7405
ChEBI CHEBI:18183
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 158°C to 162°C
Quantity 500 g
Beilstein 82132
Merck Index 14,8001
Solubility Information Soluble in water (10-15% at 20°C),alcohol,acetone,dimethyl sufoxide and methanol.
Optical Rotation −10.5° (c=5 in Water)
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
Formula Weight 129.12
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 202-700-3
RTECSNumber TW3710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only