missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylpropionic acid, 99%, Thermo Scientific™

Catalog No. p-183248
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264652500 250mg Glass Bottle
AC264650010 1g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264652500 Supplier Thermo Scientific Chemicals Supplier No. 264652500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7782-24-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00063139
InChI Key YPGCWEMNNLXISK-ZETCQYMHSA-N
Synonym s-+-2-phenylpropionic acid, s-2-phenylpropanoic acid, 2s-2-phenylpropanoic acid, s-hydratropic acid, +-hydratropic acid, s-2-phenyl-propionic acid, s-alpha-methylbenzeneacetic acid, s-2-phenylpropionic acid, s-+-hydratropic acid, unii-988109nfbc
PubChem CID 2724622
ChEBI CHEBI:48527
IUPAC Name (2S)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.1000g/mL
Boiling Point 115°C (1.0mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5223
Linear Formula CH3CH(C6H5)CO2H
Quantity 250mg
Beilstein 09, 347
Specific Gravity 1.1
Specific Rotation Condition +75° (20°C c=3,CHCl3)
Specific Rotation + 75
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Phenylpropionic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only