missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-Pentanol, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Density | 0.81 |
| Boiling Point | 118°C to 119°C |
| Flash Point | 33°C (91°F) |
| Refractive Index | 1.406 |
| Quantity | 250 mg |
| UN Number | UN1105 |
| Beilstein | 1718820 |
| Optical Rotation | +14° (Neat) |
| Formula Weight | 88.15 |
| Percent Purity | 97% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H226-H332-H335 |
| Hazard Statement | GHS H Statement H226-H332-H335 Flammable liquid and vapor. Harmful if inhaled. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS |
| RTECSNumber | SA4900000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only