missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid 98.0+%, TCI America™

Catalog No. H15685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15685G 5 g
H15681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15685G Supplier TCI America Supplier No. H15685G
Only null left

Chemical Identifiers

CAS 178306-52-0
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD11113284
InChI Key RQJWOLFMWKZKCJ-CQSZACIVSA-N
PubChem CID 15380898
IUPAC Name (2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
SMILES COC(C1=CC=CC=C1)(C2=CC=CC=C2)C(C(=O)O)O

Specifications

Melting Point 120°C
Color White
Quantity 5 g
Formula Weight 272.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid