missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Acide chloropropionique 98,0+%, TCI America™

Numéro de catalogue. C13731G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C13731G 1 g
C13735G 5 g
C137325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. C13731G Fournisseur TCI America Code fournisseur C13731G

Identifiants chimiques

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoate
SMILES C[C@H](Cl)C([O-])=O

Spécifications

Color Jaune
Quantity 1 g
UN Number 2511
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquide
Chemical Name or Material (S)-(-)-2-Chloropropionic Acid
EINECSNumber (2)-1157
RTECSNumber UA2451950
TSCA No