missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Butanol, 98+%

Catalog No. AAL1416406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1416406 5 g
AAL1416403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1416406 Supplier Thermo Scientific Chemicals Supplier No. L1416406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4221-99-2 | C4H10O | 74.123 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064281
InChI Key BTANRVKWQNVYAZ-BYPYZUCNSA-N
Synonym s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
PubChem CID 444683
ChEBI CHEBI:45475
IUPAC Name (2S)-butan-2-ol
SMILES CCC(C)O

Specifications

Density 0.806
Quantity 5 g
Flash Point 26°C (79°F)
Odor Alcohol-like
Refractive Index 1.398
UN Number UN1120
Merck Index 141541
Boiling Point 99°C to 100°C
Formula Weight 74.12
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Butanol
Show More Show Less
Hazard Category H226-H319-H335-H336
Hazard Statement GHS H Statement
H226-H319-H335-H336
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 224-168-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only