missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminononane, ChiPros 99+%, ee 99+%

Catalog No. AAL1931114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1931114 25 g
AAL1931103 1 g
AAL1931106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1931114 Supplier Thermo Scientific Chemicals Supplier No. L1931114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 869278-88-6 | C9H21N | 143.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 869278-88-6
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD03844738
InChI Key ALXIFCUEJWCQQL-UHFFFAOYNA-N
Synonym 2-nonanamine, 2-aminononane, 2-nonylamine, 2-amino-nonane, acmc-20apfg, acmc-1bh6g
PubChem CID 12878166
IUPAC Name nonan-2-amine
SMILES CCCCCCCC(C)N

Specifications

Density 0.782
Boiling Point 73°C (19 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.4271
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Optical Rotation +6° (Neat)
Formula Weight 143.27
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminononane
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 663-044-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only