missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2-Aminométhyl-1-éthylpyrrolidine 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Quantity | 1 g |
| UN Number | 2733 |
| Formula Weight | 128.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine |
Sécurité et manipulation
| EINECSNumber | (9)-0211 |
| TSCA | No |