missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Aminoethyl)isothiouronium Bromide Hydrobromide 98.0+%, TCI America™

Catalog No. A107825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A107825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A107825G Supplier TCI America Supplier No. A107825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-10-0
Molecular Formula C3H11Br2N3S
Molecular Weight (g/mol) 281.01
MDL Number MFCD00037011
InChI Key XDVMCVGTDUKDHL-UHFFFAOYSA-N
Synonym 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide
PubChem CID 5940
IUPAC Name 2-aminoethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N.Br.Br

Specifications

Melting Point 195°C
Color White
Quantity 25 g
Formula Weight 281.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-(2-Aminoethyl)isothiouronium Bromide Hydrobromide
RTECSNumber UM0175000
TSCA Yes