missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-Amino-1-propanol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 98°C |
| Quantity | 5 mL |
| UN Number | 2735 |
| Formula Weight | 75.11 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-(+)-2-Amino-1-propanol |
Safety and Handling
| EINECSNumber | (9)-0122&(2)-0323 |
| TSCA | No |