missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-phenylethanol, 97%

Catalog No. AAL1940403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1940403 1 g
AAL1940406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1940403 Supplier Thermo Scientific Chemicals Supplier No. L1940403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56613-81-1 | C8H11NO | 137.18 g/mol

(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 56613-81-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00239405
InChI Key ULSIYEODSMZIPX-MRVPVSSYSA-N
Synonym s-2-amino-1-phenylethanol, s---2-phenylglycinol, 1s-2-amino-1-phenylethanol, s-2-amino-1-phenylethan-1-ol, 1s-2-amino-1-phenylethan-1-ol, s-+-2-amino-1-phenylethanol, r---alpha-aminomethyl benzyl alcohol, pubchem13836, 2-amino-1 s-phenylethanol
PubChem CID 643217
IUPAC Name (1S)-2-amino-1-phenylethanol
SMILES NC[C@@H](O)C1=CC=CC=C1

Specifications

Melting Point 57°C to 63°C
Boiling Point 160°C (17 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 2412760
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Optical Rotation +42° (c=2 in Ethanol)
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Amino-1-phenylethanol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only