missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine 99.0+%, TCI America™

Catalog No. M09851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09851G Supplier TCI America Supplier No. M09851G
Only null left

Specifications

CAS 84249-39-8
Melting Point 83°C
Color Yellow
Molecular Formula C13H13N3O2
Quantity 1 g
MDL Number MFCD00145247
InChI Key RAKMWGKBCDCUDX-JTQLQIEISA-N
SMILES CC(C1=CC=CC=C1)NC2=NC=C(C=C2)[N+](=O)[O-]
IUPAC Name 5-nitro-N-[(1S)-1-phenylethyl]pyridin-2-amine
Molecular Weight (g/mol) 243.266
PubChem CID 29772754
Formula Weight 243.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-(alpha-Methylbenzylamino)-5-nitropyridine
Show More Show Less