missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Acetoxypropionic Acid 98.0+%, TCI America™

Numéro de catalogue. A12995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A12995G 5 g
A129925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A12995G Fournisseur TCI America Code fournisseur A12995G
Il en reste null

Identifiants chimiques

CAS 6034-46-4
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00191370
InChI Key WTLNOANVTIKPEE-VKHMYHEASA-N
Synonym (-)-O-Acetyl-L-lactic Acid
PubChem CID 6326324
IUPAC Name (2S)-2-acetyloxypropanoic acid
SMILES CC(C(=O)O)OC(=O)C

Spécifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 132.12
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Acetoxypropionic Acid
TSCA No
Recommended Storage Refrigerator